Oakton pH 150 Waterproof Portable Meter, Cole Palmer
Supplier: Antyila Scientific
35614-30 35614-32
76221-258EA 1533.42 CAD
76221-258 76221-260
Oakton pH 150 Waterproof Portable Meter, Cole Palmer
Electrochemistry Meters pH and Multi-Parameter Meters
Oakton pH 150 waterproof portable meter is rugged, accuratea and reliable and an IP67-rated waterproof meter.
  • Intuitive, user-friendly icons-electrode status displays pH slope
  • Expanded data logging for up to 150 data sets
  • mV offset for calibration of ORP standards, zeroing mV value, and pH diagnostics
  • 700 hours battery-life or optional universal power adapter
Grip-Clip™ holder quickly and easily secures the electrode to your beaker or container. The built-in stand and wall mount option allow for easy benchtop measurement or storage.
Delivery information: Supplied with Grip-Clip electrode holder and two AA batteries.