Exo-Terra® Ceramic Heater
Supplier: ROLF C. HAGEN (USA) CORP.
Exo-Terra®
PT2048 PT2046 PT2044 PT2047 PT2045
470347-694EA 63.4 CAD
470347-694 470347-860 470347-856 470308-862 470347-842
Exo-Terra® Ceramic Heater
Animal Supplies Animal Environmental Enrichment
Radiant heat without light.
  • 99% heat efficiency
  • Emits a natural "Sun-Like" infrared heat
  • Does not disturb normal photo periods
  • Solid ceramic element, suitable for humid environments
The Exo Terra Ceramic Heater is a radiant heat source which creates a natural sun-like heat with a uniform pattern. The long wave infrared heat produced by this emitter will warm reptiles very efficiently. It also increases the ambient air-temperature in the terrarium.
Infrared heat penetrates the scales and skin tissue, promoting health and healing as it widens the blood vessels and increases blood circulation. The Ceramic Heater is to be used only with the specially designed Exo Terra Wire Light.