Adhesive Paper Labels 'Exempt Specimen', Labelmaster
Supplier: Label Master
L358 L361
101377-351EA 77.53 CAD
101377-351 101377-353
Adhesive Paper Labels 'Exempt Specimen', Labelmaster
Labels
These paper labels can be used for shipping medical specimens.
Comply with the requirements in International Air Transport, ICAO and DOT for marking the package orientation. The bright red label surface has chemical and abrasion resistance, and the backing offers strong adhesion to paint, steel, fiber and plastic surfaces.