Marina® Air Pumps
Supplier: Pet World Inc
11116 11110 11112 11118
470007-538EA 35.45 CAD
470007-538 470102-116 470150-594 470308-584 470308-586
Marina® Air Pumps
Animal Supplies Animal Environmental Enrichment
Proper aeration is a must.
  • Single output pumps
  • Quiet operation
The Marina air pumps combines high performance output with quiet operation. A reliable source of air for increasing water movement and turbulence for proper oxygenation. They can also be used to operate items such as air stones, air curtains and air-actuated ornaments. Each has a single outlet.
Indicated size for each pump is the recommended Maximum tank size. If you plan on running multiple airlines (air stones, ornaments filters etc.), you should choose a larger air pump.