This bottle is excellent for near off-shore (to 200 m) trace metal sampling, since there are no metal parts to touch the sample.
- No metal parts to touch the sample
- Designed for trace metal sampling
- May be used for organic sampling to concentrations of mg/L
Intended for shallow or deep waters, these bottles are called "horizontal" because they descend parallel to the bottom. They are pulled sideways just before closing by the operator, which ensures a representative water sample for that specific depth. Bottles are ideal for sampling at the thermocline, at other stratification levels, or just above the bottom sediments. These popular and versatile bottles come in a choice of transparent acrylic or opaque PVC. PVC is more durable and less costly, but the acrylic allows viewing of the bottle contents immediately.
When running metal, organic, or any other chemical analyses, it is best to run a blank on the sampler prior to use. A thermometer can be mounted inside acrylic Beta bottles.
Delivery information: Includes bottle, carry case, messenger, and 100' line.
Caution: Volume may vary slightly. It is recommended that users verify the exact volume of the bottle prior to use.