Cole-Parmer® Deuterium Lamps
Supplier: Cole-Parmer
1194111 1194116 1194123 11941-10 11941-12 11941-13 11941-14 11941-17 11941-19 11941-20 11941-21 11941-26 11941-27 11941-28 11941-29 11941-31 11941-33 11941-34 11941-35 11941-36 11941-11 11941-16 11958-00
CA76181-236EA 2642.79 CAD
CA76181-236 CA76181-238 CA76181-240 CA76181-242 76182-480 76182-482 76182-484 76182-486 76182-488 76182-490 76182-494 76182-496 76182-498 76182-502 76182-504 76182-506 76182-508 76182-510 76182-616 76182-618 76182-620 76182-622 76182-624 76182-626 76182-628 76473-744 76473-754 76473-760 76473-762 76473-764 76473-766 76473-784
Cole-Parmer® Deuterium Lamps
Lights Deuterium Lamps
Economical replacement deuterium lamps.
  • Select from standard or long life lamps
  • Manufactured in an ISO accredited facility
  • Individually tested and issued with a Test Certificate
This range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. The lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
These deuterium lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable fromMore Product Information