DuPont™ ProClean® 3 Sterile Procedure Gowns with Welded Seams
Supplier: Avantor
76290-272CS 395.64 CAD
76290-272
DuPont™ ProClean® 3 Sterile Procedure Gowns with Welded Seams
Disposable Garments
DuPont™ ProClean® 3 Sterile Procedure Gowns are made from spunbond meltblown spunbond (SMS) fabric with anti-static treatment.
  • Sterile
  • Anti-static
  • Welded seams
  • Repels alcohol
  • Low lint generation
  • Set in sleeves
  • Extra-long hook and loop closure at neck
Sterile procedure gowns are low lint generating and alcohol repellent. These gowns also feature set in sleeves, generous cut, and an extra-long hook & loop closure at the neck for a customized fit.
Certifications: Ethylene Oxide 10-6 Sterility Assurance Level. Meets ANSI/AAMI PB70:2012 Level III Performance Requirements. Meets 16 CFR Part 1610 Class 1 Flammability Rating. Thickness meets ASTM D3776.
Packaging: Sterile gowns are individually packaged with 32 gowns per case.