Dulbecco's Phosphate-Buffered Saline (DPBS), Himedia®
Supplier: HIMEDIA LABORATORIES PVT. CA
TS1006-5L
TS1006-20L
CA95016-726EA
126.02
CAD
CA95016-726
CA95016-728
CA95016-730
95016-726
Dulbecco's Phosphate-Buffered Saline (DPBS), Himedia®
Buffers
Cell Culture Buffers
Dulbecco's phosphate buffered saline is used for tissue disaggregation and monolayer dispersal.The presence of calcium and magnesium ions hinder the trypsin activity.
- Off-white to creamish white
- pH: 7.20 to 7.80
- Osmolality without sodium bicarbonate: 275.00 to 315.00 mOsm
- Endotoxin content: NMT 1 EU/ml
The solution is used to maintain the medium within physiological pH range and intracellular and extra cellular osmotic balance. Modified with a carbohydrate, such as glucose serves as an energy source for cell metabolism.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...