Bushnell Haas Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M349-500G M349-2.5KG M349-5KG
CA61000-080EA 1673.99 CAD
CA61000-080 CA89225-252 CA89225-254
Bushnell Haas Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Bushnell Haas Agar are recommended for the microbial examination of fuels and for studying microbial hydrocarbon deterioration.
  • White to cream homogeneous free flowing powder
  • Firm gelling, comparable with 2.0% agar gel
  • pH: 6.80 to 7.20
  • Store below 30 °C
Bushnell Haas Agar contains magnesium sulfate (0.200 g/l); calcium chloride (0.020 g/l); monopotassium phosphate (1.000 g/l); dipotassium phosphate (1.000 g/l); ammonium nitrate (1.000 g/l); ferric chloride (0.050 g/l) and agar (20.000 g/l).