AMPLATE™ PCR Reaction Plates, Chemglass
Supplier: Chemglass
CLS-3615-24B CLS-3615-24G CLS-3615-24N CLS-3615-24R CLS-3615-24Y CLS-3615-48B CLS-3615-48G CLS-3615-48N CLS-3615-48R CLS-3615-48Y
89492-962CS 122.5 CAD
89492-962 89492-964 89492-966 89492-968 89492-970 89492-972 89492-974 89492-976 89492-978 89492-980
AMPLATE™ PCR Reaction Plates, Chemglass
Microplates PCR Plates
These PCR plates are thin-walled and designed for rapid thermal transfer.
  • Pre-cut plates
  • Each well has a capacity of 0.2 ml
They are precision molded to ensure well-to-well and plate-to-plate uniformity. The insides of the tubes are smooth and have an inert surface which enzymes and nucleic acids do not bind.
The plates will accommodate differences in expansion coefficients between the metal thermal cycler block and polypropylene tubes. To facilitate orientation, the bottom right corner is cut away.