Calbiochem® Phosphatase Inhibitor Cocktail Set III, EMD Millipore
Supplier: MilliporeSigma
524627-1ML 524627-1SET
CA80017-332EA 618.15 CAD
CA80017-332 CA80017-334
Calbiochem® Phosphatase Inhibitor Cocktail Set III, EMD Millipore
Cell Culture Supplements and Additives
A cocktail of four phosphatase inhibitors for broad-spectrum inhibition of both serine/threonine and protein tyrosine phosphatases.
  • Toxic
Caution: For long-term storage, aliquot and freeze (-20 °C). Dilute 1:5 just prior to use.