Goldenrod Animal Lancet, Braintree Scientific
Supplier: BRAINTREE SCIENTIFIC INC MS
GR-4MM GR-5MM
MSPP-GR4MMCS 130.77 CAD
MSPP-GR4MM MSPP-GR5MM
Goldenrod Animal Lancet, Braintree Scientific
Blood Lancets
Animal lancet for blood collection of facial vein (submandibular) blood samples.
  • Sterile, disposable blood lancet
  • Inexpensive, easy, and fast to use
Multiple blood samples can be taken from small animals without hurting the animals.
Packaging: Packed in 5 trays of 200.