KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Screw Cap, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60941A-8 60941A-12 60941A-16 60941A-40 60941A-24
11310-768CS 305.42 CAD
11310-768 11310-770 11310-772 11310-776 89003-542
KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Screw Cap, Kimble Chase, DWK Life Sciences
Vials
Clear, screw thread sample vials.
  • Black phenolic cap and cone-shaped polyethylene liner
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Material conforming to USP Type I and ASTM E438, Type I, Class A requirements.