Versilic® Peroxide Cured Silicone Stoppers, Saint-Gobain

Supplier: Saint Gobain Performance Plastics
DX263008-50 DX263021-20 DX263034-05 DX263038-05 DX263045-05 DX263075-02 DX263100-01 DX263006-50 DX263013-50 DX263015-50 DX263054-02 DX263070-02 DX263092-01
66111-248PK 58.02 CAD
66111-248 66111-260 66111-270 66111-272 66111-276 66111-288 66111-294 66111-296 66111-246 66111-252 66111-254 66111-280 66111-286 66111-292
Versilic® Peroxide Cured Silicone Stoppers, Saint-Gobain
Stoppers Silicone Stoppers
Highly recommended for all types of tissue culture work.

  • Non-toxic
  • Chemically and biologically neutral
  • Temperature up to 250 °C (482 °F)

They are suitable for use at temperatures between –70 to 250 °C (–94 to 482 °F).

These stoppers are odorless, tasteless, non-adherent and resistant to aging.

They are ideal in biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus.

Certifications: DIN Standard 18271, FDA/BGA approved.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR