Joints, O-ring seals, Chemglass
Supplier: Chemglass
CG-124-09 CG-124-10 CG-124-01 CG-124-02 CG-124-03 CG-124-04 CG-124-05 CG-124-06 CG-124-07 CG-124-08 CG-124-20
80073-184EA 144.73 CAD
80073-184 80073-186 80073-168 80073-170 80073-172 80073-174 80073-176 80073-178 80073-180 80073-182 80073-188
Joints, O-ring seals, Chemglass
NS Joint Sleeves and Connectors Joint Sleeves
Joint has an O-ring groove to accept the various O-ring size.
Provides for a grease-free, vacuum seal when used with CG-150 clamps.
Delivery information: Each joint is supplied with a Viton O-ring of the listed size.