VWR® Sleek Safety Glasses

Supplier: VWR
VWR®
PS-25
76210-826CS 2146.37 CAD
76210-826
VWR® Sleek Safety Glasses
Safety Glasses
Lightweight and sleekly designed safety glasses.

  • Super lightweight at 22.7 g
  • Dielectric construction
  • Comfortable nose piece
  • Slim temple with rubber pad
  • Impact-resistant
  • Polycarbonate lens
  • Provides 99.9% UV protection

Certifications: Meets ANSI Z87.1 standards.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR