2-Propanol ≥70% (v/v)

Supplier: INNOVA CHEMICALS CA
Danger

INN890551 INN890552
CA76288-096EA 410 CAD
CA76288-096 CA76288-098
2-Propanol ≥70% (v/v)
2-Propanol

For laboratory cleaning and disinfection purposes. Non-sterile.

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (179.6 °F)
Melting Pt: –88 °C / –126.4 °F
Density: 0.8…0.92 g/cm³ (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1987
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form/Colour liquid/colourless

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR