Water, deionized

Supplier: INNOVA CHEMICALS CA

Synonyms: Hydrogen oxide

INN890546
CA76288-086EA 133.38 CAD
CA76288-086
Water, deionized
Water

This general-purpose water is purified by deionization. Water has a pH of 5.0 to 7.0. It passes the test for reducing potassium permanganate.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 1.000 g/cm³ (3.98 °C)
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Form/Colour liquid/colourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR