Sodium hydroxide ACS

Supplier: INNOVA CHEMICALS CA
Danger

Synonyms: Caustic soda

INN890543
CA76288-080EA 826.86 CAD
CA76288-080
Sodium hydroxide ACS
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Merck Index: 14,08627

Order Now

Specification Test Results

Form/Colour powder/white

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR