Sodium hexametaphosphate, Lab-Grade

Supplier: INNOVA CHEMICALS CA

Synonyms: Sodium metaphosphateSHMP

INN890547
CA76288-088EA 2171.25 CAD
CA76288-088
Sodium hexametaphosphate, Lab-Grade
Sodium hexametaphosphate
Formula: Na₆P₆O₁₈
MW: 611.77 g/mol
Melting Pt: 610 °C
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00136045
CAS Number: 10124-56-8

Order Now

Specification Test Results

Form/Colour solid/white

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR