Calcium chloride dihydrate ACS
Supplier: INNOVA CHEMICALS CA
|
Warning
|
INN890519
INN890520
CA76288-566CS
115
CAD
CA76288-566
CA76288-568
Calcium chloride dihydrate ACS
Calcium chloride dihydrate
Formula:
CaCl₂·2H₂O Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 176 °C Density: 1.85 g/cm³ (20 °C) |
MDL Number:
MFCD00149613 CAS Number: 10035-04-8 Merck Index: 13,01660 |
Specification Test Results
Form/Colour | crystalline/white |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...