EDTA disodium salt dihydrate, powder ACS

Supplier: INNOVA CHEMICALS CA
Warning

INN890512 INN890513
CA76288-458EA 1883.94 CAD
CA76288-458 CA76288-694 CA76288-696
EDTA disodium salt dihydrate, powder ACS
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 248 °C
MDL Number: MFCD00150037
CAS Number: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form/Colour solid/colourless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR