Urea, crystals ACS

Supplier: INNOVA CHEMICALS CA

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

INN890516 INN890517 INN890518
CA76288-460EA 76 CAD
CA76288-460 CA76288-562 CA76288-564
Urea, crystals ACS
Urea
Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00008022
CAS Number: 57-13-6
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form/Colour solid/white


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR