Titration solvent ASTM D-664

Supplier: INNOVA CHEMICALS CA
Danger

INN890557
CA76288-108CS 373.76 CAD
CA76288-108
Titration solvent ASTM D-664
Titration solvent
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
CAS Number: 108-88-3
UN: 1294
ADR: 3,II

Order Now

Specification Test Results

Form/Colour liquid/colourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR