Ethanol denatured, anhydrous (MeOH + Ethylacetate unspecified)

Supplier: INNOVA CHEMICALS CA
Danger

Synonyms: Ethyl alcohol denatured

INN890553 INN890554 INN890555
CA76288-100CS 404 CAD
CA76288-100 CA76288-102 CA76288-104
Ethanol denatured, anhydrous (MeOH + Ethylacetate unspecified)
Ethanol denatured

Ethyl alcohol (anhydrous) 85.5% v/v (approx.), methanol 13.7% v/v (approx.), ethyl acetate 0.85% v/v (approx.).

Formula: H₃CCH₂OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II

Order Now

Specification Test Results

Form/Colour liquid/colourless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR