Ethanol denatured ACS (denatured with methanol and ethyl acetate)

Supplier: INNOVA CHEMICALS CA
Danger

Synonyms: Ethyl alcohol denatured

INN890542
CA76288-078EA 296.7 CAD
CA76288-078
Ethanol denatured ACS (denatured with methanol and ethyl acetate)
Ethanol denatured

Reagent Alcohol ACS (denatured with methanol and ethyl acetate).
Contains approx. 95% Ethyl alcohol, 13.7% v/v Methanol , ethyl acetate 0.85% v/v as denaturing agents.

Formula: H₃CCH₂OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Form/Colour liquid/colourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR