Tech-Med® Instrument Trays, DUKAL Corporation®
Supplier: Dukal
4274 4276
76235-314EA 89.29 CAD
76235-314 76235-318
Tech-Med® Instrument Trays, DUKAL Corporation®
Trays/Dishes/Pots Instrument Trays/Dishes
Trays are crafted without seams to prevent bacteria growth and allow for easy cleaning.
  • Made of the highest grade stainless steel (18/8)
  • Autoclavable