AquaClear® Power Filters
Supplier: ROLF C. HAGEN (USA) CORP.
A620 A613 A606 A1372 A614 A601 A1373 A605 A1374 A597 A622 A599 A611 A600 A1371 A624 A615 A602 A618 A612 A619 A616 A617 A596 A598 A1370 A623 A610 A595
470308-656EA 6.5 CAD
470308-656 470308-634 470308-678 470308-756 470308-658 470308-636 470308-758 470308-638 470308-670 470308-672 470308-650 470308-750 470308-652 470308-752 470308-676 470308-654 470308-632 470308-754 470308-680 470308-646 470308-668 470308-648 470308-660 470308-640 470308-662 470308-642 470308-664 470308-644 470308-666
AquaClear® Power Filters
Animal Supplies Animal Environmental Enrichment
The AquaClear Power Filters features a unique multi-stage filtration system that provides complete mechanical, chemical and biological filtration for superior water quality. The Power Filter has a filtration volume that is up to seven times larger than comparable filters. It also allows superior contact time with filter media. The filter preserves beneficial bacterial creating a stress-free environment for aquarium inhabitants and maintaining clear aquarium water.
  • Easy operation and maintenance
  • Large capacity cartridges out last competitors
  • Easy to use tank filtration
Its unique waterfall design enables filtered water to return to the tank silently, gently breaking the surface tension and creating beneficial oxygenation. The Power Filters patented flow control feature allows you to adjust the water flow according to your aquarium needs. The media basket prevents water from by-passing the filter media, maximizing filtration efficiency. Installation and maintenance are quick, easy and convenient. The filter comes equipped with AquaClear Foam, Activated Carbon and BioMax.