Empty Optimus Racks for CleanPak™ and TipEject™ Pipette Tip Reloads, Biotix
Supplier: Biotix
63300151 63300152
76266-138CS 96.83 CAD
76266-138 76266-140
Empty Optimus Racks for CleanPak™ and TipEject™ Pipette Tip Reloads, Biotix
Pipette Tip Boxes and Racks
The Optimus rack is compatible with CleanPak™ and TipEject™ reloads for easy transfer and storage.
  • Double ball hinge allows the lid to open 180° for optimal multichannel pipetting
  • Ribs in the base & snapcard interact to reduce snapcard flex for consistent liquid levels with multichannel devices
  • Withstands repeatable autoclaving and reuse
  • Locking clasp secures tips during transport and non-use
  • Great stackability for easy storage
The Optimus rack is compatible with both uTIP™ and xTIP4™ pipette tip lines. The empty racks are available in two sizes; for tip volumes from 10 to 300 µl or 1000 to 1250 µl. Both racks are packaged 10 per pack, 4 packs per case.