L(+)-Tartaric acid ACS

Supplier: Ricca Chemical
Danger

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

RDCT0030-125B1 RDCT0030-500B1
RCRDCT003125B1EA 94.8 CAD
RCRDCT003125B1 RCRDCT003500B1
L(+)-Tartaric acid ACS
L(+)-Tartaric acid
Formula: C₄H₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min 99.0 %
Heavy Metals (as Pb) max 5 ppm
Oxalate (C₂O₄) To Pass Test
Phosphate (PO₄) max 0.001 %
Sulfur compounds (as SO₄) max 0.002 %
Chloride (Cl) max 0.001 %
Iron (Fe) max 5 ppm
Insoluble Matter max 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR