Sulfuric acid ACS

Supplier: Ricca Chemical
Danger

RABS0030-1C RABS0030-4A RABS0030-4C RABS0030-2.5D RABS0030-20E1 RABS0030-500C RABS0030-2.5D1
RCRABS0030-1CEA 216.46 CAD
RCRABS0030-1C RCRABS0030-4A RCRABS0030-4C RCRABS00302.5D RCRABS003020E1 RCRABS0030500C RCRABS0032.5D1
Sulfuric acid ACS
Sulfuric acid
Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay 95.0-98.0 %
Color (APHA) max 10
Ammonium (NH₄) max 2 ppm
Heavy Metals (as Pb) max 1 ppm
Nitrate (NO₃) max 0.5 ppm
Chloride (Cl) max 0.2 ppm
Iron (Fe) max 0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR