Orthophosphoric acid ACS
Supplier: Ricca Chemical
|
Danger
|
RABP0030-1C
RABP0030-4C
RABP0030-2.5D1
RABP0030-500D1
RCRABP0030-1CEA
203.82
CAD
RCRABP0030-1C
RCRABP0030-4C
RCRABP0032.5D1
RCRABP003500D1
Orthophosphoric acid ACS
Orthophosphoric acid
Formula:
H₃PO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 UN: 1805 ADR: 8,III |
Specification Test Results
Assay | min 85.0 % |
Color (APHA) | max 10 |
Heavy Metals (as Pb) | max 0.001 % |
Nitrate (NO₃) | max 5 ppm |
Sulfate (SO₄) | max 0.003 % |
Sodium (Na) | max 0.025 % |
Magnesium (Mg) | max 0.002 % |
Chloride (Cl) | max 3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...