Nitric acid 30% (v/v) in methanol
Supplier: Ricca Chemical
|
Danger
|
R5320300-1A
R5320300-1C1
RCR5320300-1AEA
372.88
CAD
RCR5320300-1A
RCR5320300-1C1
Nitric acid 30% (v/v) in methanol
Nitric acid
Formula:
HNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 UN: 2924 ADR: 3 (8),II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sodium Hydroxide/Phenolphthalein) | 29.8-30.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...