Hydrochloric acid 20% (v/v)

Supplier: Ricca Chemical
Danger

R3574000-4A R3574000-500A R3574000-1A R3574000-10F R3574000-20F
RC3574-1EA 235.42 CAD
RC3574-1 RC357416 RC357432 RCR3574000-10F RCR3574000-20F
Hydrochloric acid 20% (v/v)
Hydrochloric acid
Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to slightly greenish-yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 19.92-20.08 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR