Formic acid 2% (w/v)

Supplier: Ricca Chemical
Danger

Synonyms: Aminic Acid, Methanoic acid

R3213000-10F R3213000-20F R3213000-4A
RCR3213000-10FEA 243.32 CAD
RCR3213000-10F RCR3213000-20F RCR3213000-4A
Formic acid 2% (w/v)
Formic acid
Formula: HCO₂H
MW: 46.03
Density: 1.22
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 1.99-2.01 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR