Citric acid 10% (w/v), from USP materials

Supplier: Ricca Chemical
Danger

R2120000-4A R2120000-500A
RCR2120000-4AEA 297.04 CAD
RCR2120000-4A RCR2120000500A
Citric acid 10% (w/v), from USP materials
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 9.97-10.03 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR