Chromic acid 10% (w/v)
Supplier: Ricca Chemical
|
Danger
|
R2077000-4C
R2077000-500C
RCR2077000-4CEA
440.82
CAD
RCR2077000-4C
RCR2077000500C
Chromic acid 10% (w/v)
Chromic acid
Formula:
CrH₂O₄ Storage Temperature: Ambient |
CAS Number:
7738-94-5 UN: 1755 ADR: 8,II |
Specification Test Results
Appearance | Orange liquid |
Assay (vs. Sodium Thiosulfate/Starch) | 9.9-10.1 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...