L(+)-Ascorbic acid ACS

Supplier: Ricca Chemical

RDCA0750-100B1 RDCA0750-500B1
RCRDCA075100B1EA 306.52 CAD
RCRDCA075100B1 RCRDCA075500B1
L(+)-Ascorbic acid ACS
L(+)-Ascorbic acid
Formula: C₆H₈O₆
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min 99.0 %
Specific rotation 20.5-21.5 °
Heavy Metals (as Pb) max 0.002 %
Iron (Fe) max 0.001 %
Residue after Ignition max 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR