L(+)-Ascorbic acid 2% (w/v)
Supplier: Ricca Chemical
Synonyms:
Ascorbic Acid
, L-Ascorbic acid
, Acidum ascorbicum
, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one
R0808200-1C
RCR0808200-1CEA
439.1
CAD
RCR0808200-1C
L(+)-Ascorbic acid 2% (w/v)
L(+)-Ascorbic acid
Formula:
C₆H₈O₆ MW: 176.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064328 CAS Number: 50-81-7 |
Specification Test Results
Appearance | Colorless to slightly yellow liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...