Platinum, disc, drilled apertures 6 mm O.D., Thickness: 0.24 mm

Supplier: Electron Microscopy Sciences

Synonyms: Platinium

46050-PT 46100-PT 46020-PT 46030-PT
100496-756EA 373.29 CAD
100496-756 100496-758 100496-752 100496-754
Platinum, disc, drilled apertures 6 mm O.D., Thickness: 0.24 mm
Platinum

Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs.


Platinum Apertures size tolerances vary as follow:
• Nominal hole diameter: +/- 1 micron
• Disc diameter: +/- 10 microns
• Disc thickness: +/- 10 microns


Platinum Apertures size tolerances vary as follow:


Hole Sizes not listed are available upon special request.

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR