Kimble® Accuform® Aluminum Seal Microvials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60720-3 60720-5 60720-1 60720-2 60730-310
66010-636PK 341.5 CAD
66010-636 66010-637 66010-634 66010-635 66010-642
Kimble® Accuform® Aluminum Seal Microvials, DWK Life Sciences
Vials
Accuform® microvials feature a V-shaped interior to enable recovery of a minute residual sample by means of a syringe needle.
  • Designed for an aluminum seal
  • Graduated and ungraduated
  • Closures not included
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.