X-Ray Mix Additive, Chemplex®, tablets
Supplier: CHEMPLEX INDUSTRIES
625
650
15566-075EA
426.21
CAD
15566-075
15566-077
X-Ray Mix Additive, Chemplex®, tablets
X-Ray Mix Additive, Chemplex®
Highly “plasticizes” and firmly bonds samples together under briquetting pressure
Produces sample pellets that are characteristically smooth, unblemished, glossy and highly resistant to breakage. General purpose additive applicable to diversified sample material preparations. Water soluble.