Magid® EconoWear® Poly Plus® Two Layer Polypropylene Lab Coat, Magid Glove
Supplier: Magid Glove
C9PXL C9-P-L C9-P-M
69100-134CS 833.56 CAD
69100-134 32893-128 32893-130
Magid® EconoWear® Poly Plus® Two Layer Polypropylene Lab Coat, Magid Glove
Disposable Garments
Disposable Lab Coats are made for jobs specifically requiring tough, economical, liquid repellent apparel.
  • Provides increased barrier protection from particulates
  • Features a snap front closure for comfortable fit
This material is laminated to a layer of white polypropylene film which produces a super strong garment that resists snags and punctures, while providing increased barrier protection from particulates.
Packaging: 25 per case.