BioReady Taq Polymerase, Bulldog Bio
Supplier: Bulldog Bio
BSA12L020 BSA12L050
102408-082EA 207.69 CAD
102408-082 102408-084
BioReady Taq Polymerase, Bulldog Bio
Nucleic Acid Reagents End-point PCR Enzymes and Kits
High Quality Recombinant PCR Enzyme.
Cost effective – BioReady Taq Polymerase is a great choice for your everyday PCR needs. This recombinant DNA polymerase is optimized for improved efficiency and can amplify between 50 bp and 5000 bp.
BioReady Taq is manufactured under stringent GMP control, and though we don’t recommend it, this ultra-pure enzyme extract is still fully active after days at 37 °C.