GFP-Traps, Bulldog Bio
Supplier: Bulldog Bio
GTA010 GTA200 GTA400 GTAK020
102408-032EA 586.15 CAD
102408-032 102408-038 102408-040 102408-042
GFP-Traps, Bulldog Bio
Protein Linking and Labeling Systems
Novel Antibody Fragment that binds GFP Fusion Proteins.
GFP-Traps utilize super-high affinity Camelidae antibody fragments coupled to agarose beads or magnetic agarose beads. These “Nanobody-Traps” are perfect for immuno-precipitation, immuno-purification and immuno-pull down experiments with up to 10-fold better purity and yield than that of classic mouse monoclonal antibodies.
Compatible with a variety of source materials, Nanobody-Traps may be used with mammalian cells, tissues & organs, bacteria, yeast and even plants. These reagents allow your GFP-fusions to be perfect candidates for immunoprecipitations, Co-IP, mass spectroscopy, enzyme activity measurements and ChIP analysis.