Mapa® Flock-Lined Z Pattern Nitrile Gloves, Magid
Supplier: Magid Glove
MF18-10 MF18-11 MF18-9 MF187
32893-146CS 533.53 CAD
32893-146 32893-148 32893-150 69100-156 69100-158
Mapa® Flock-Lined Z Pattern Nitrile Gloves, Magid
Gloves
Flock-lined 100% nitrile glove.
  • High puncture resistance
  • Chemical resistant and z-pattern grip
Tough and versatile enough to handle any job that comes their way. This style is also USDA accepted for use in food processing settings.