Interleukin-1β (Human) ELISA Kit, Cayman Chemical

Supplier: Cayman Chemical Company

583311-480 583311-96
75816-956EA 3092.46 CAD
75816-956 75816-958
Interleukin-1β (Human) ELISA Kit, Cayman Chemical
Assays ELISAs

An immunometric assay for the measurement of IL-1β.


  • Measure IL-1β from human plasma, serum, synovial fluid, and other sample matrices
  • Incubation: 18 hours
  • Development: 60 to 90 minutes
  • Read: Colorimetric at 405 to 420 nm


Cayman’s IL-1β (human) ELISA is an immunometric (i.e., sandwich) ELISA that permits IL-1β measurements within the range of 3.9-250 pg/ml, typically with a limit of detection of 3.9 pg/ml.

Order Now

SPECIFICATIONS

Environmentally Preferable
Primary antibody reactivity Human
Assay Type Sandwich
Host

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR