Magid® Softside Indirect Cap Vent Safety Goggles, Magid Glove
Supplier: Magid Glove
151 151FF
66200-102CS 1127.2 CAD
66200-102 66200-104
Magid® Softside Indirect Cap Vent Safety Goggles, Magid Glove
Goggles
Magid® softside indirect cap safety goggles feature with a clear, impact-resistant lens and an indirect cap-vented frame for exceptional eye protection and splash resistance.
  • Availabel with standard and fog free lens
These economically priced safety goggles feature with a soft construction that delivers a comfortable fit and allows for good peripheral vision.
Certifications: Meets ANSI Z87.1-2010 High-Impact Standards.
Packaging: Each pair is individually packed in a sealed polybag.
Order Now
SPECIFICATIONS
Environmentally Preferable