Anti-GlcUAbeta1-3GalNAc(4S) Mouse Monoclonal Antibody [clone: LY111]

Supplier: TCI
A3143-VIAL
TCA3143-VIALEA 976.44 CAD
TCA3143-VIAL
Anti-GlcUAbeta1-3GalNAc(4S) Mouse Monoclonal Antibody [clone: LY111]
Antibodies
The monoclonal antibodies preferentially recognize chondroitin sulfate containing the GlcUAβ1-3GalNAc(4S) disaccharide unit (A-unit). No cross-react with hyaluronic acid, dermatan sulfate, keratan sulfate and heparan sulfate.

Type: Primary
Antigen: GlcUAbeta1-3GalNAc(4S)
Clonality: Monoclonal
Clone: LY111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR