Anti-GlcUA(2-O-sulfate) beta1-3GalNAc(6-O-sulfate) Mouse Monoclonal Antibody [clone: MO-225]

Supplier: TCI
A2872-VIAL
TCA2872-VIALEA 726.8 CAD
TCA2872-VIAL
Anti-GlcUA(2-O-sulfate) beta1-3GalNAc(6-O-sulfate) Mouse Monoclonal Antibody [clone: MO-225]
Antibodies
The monoclonal antibodies preferentially recognize chondroitin sulfate containing the GlcUA(2-O-sulfate) β1-3GalNAc(6-O-sulfate) disaccharide unit (D-unit). No cross-react with hyaluronic acid, dermatan sulfate, keratan sulfate and heparan sulfate.

Type: Primary
Antigen: GlcUA(2-O-sulfate) beta1-3GalNAc(6-O-sulfate)
Clonality: Monoclonal
Clone: MO-225
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR